Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methoxy-5-sulfamoylbenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B23698-09 B23698-18 B23698-09 B23698-18 B23698-30
AAB23698-09EA 53.89 USD InStock
AAB23698-09 AAB23698-18 CAAAB23698-18 CAAAB23698-09 AAB23698-30
2-Methoxy-5-sulfamoylbenzoic acid 97%
2-Methoxy-5-sulphamoylbenzoic acid


Formula: C₈H₉NO₅S
MW: 232.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129997
CAS Number: 22117-85-7
EINECS: 244-789-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty