Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N'-Diphenyl-p-phenylenediamine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N,N'-Diphenyl-1,4-phenylenediamine

B23713-06 B23713-14 B23713-22 B23713-22 B23713-06 B23713-14
AAB23713-14EA 100.99 USD InStock
CAAAB23713-06 CAAAB23713-14 AAB23713-14 AAB23713-06 CAAAB23713-22 AAB23713-22
N,N'-Diphenyl-p-phenylenediamine 97%
N,N'-Diphenyl-p-phenylenediamine


Formula: C₁₈H₁₆N₂
MW: 260.34 g/mol
Boiling Pt: 220…225 °C/1 mm
Melting Pt: 146…153 °C
Density: 1.28
Storage Temperature: Ambient
MDL Number: MFCD00003015
CAS Number: 74-31-7
EINECS: 200-806-4
Merck Index: 12,03388
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty