Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-[(Trifluoromethyl)thio]aniline 96%

Supplier: Thermo Scientific Chemicals
Warning

B23734-03 B23734-03 B23734-06 B23734-06
AAB23734-06EA 353.99 USD InStock
CAAAB23734-03 CAAAB23734-06 AAB23734-06 AAB23734-03
3-[(Trifluoromethyl)thio]aniline 96%
3-[(Trifluoromethyl)thio]aniline


Formula: C₇H₆F₃NS
MW: 193.19
Boiling Pt: 221…224 °C
Density: 1.348
Flash Pt: 103 °C(218 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190130
CAS Number: 369-68-6
EINECS: 206-722-4
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty