Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethylbutyryl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

B23774-22 B23774-14 B23774-14 B23774-22
AAB23774-14EA 115.99 USD InStock
AAB23774-14 AAB23774-22 CAAAB23774-14 CAAAB23774-22
2-Ethylbutyryl chloride 99%
2-Ethylbutyryl chloride


Formula: C₆H₁₁ClO
Boiling Pt: 137…139 °C
Density: 0.980
Flash Pt: 33 °C(91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00018812
CAS Number: 2736-40-5
EINECS: 220-359-9
UN: 2920
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty