Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-phenylalanine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Ac-Phe-OH

B23812-03 B23812-14 B23812-03 B23812-06 B23812-06 B23812-14
AAB23812-06EA 72.99 USD InStock
AAB23812-06 AAB23812-14 AAB23812-03 CAAAB23812-06 CAAAB23812-03 CAAAB23812-14
N-Acetyl-L-phenylalanine 99%
N-Acetyl-L-phenylalanine


Formula: C₁₁H₁₃NO₃
MW: 207.2 g/mol
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00063158
CAS Number: 2018-61-3
EINECS: 217-959-8
Order Now
Specification Test Results
Optical rotation +40° (c=1 in methanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty