Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Cyanobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

B23836-06 B23836-06 B23836-22 B23836-14 B23836-14 B23836-22
AAB23836-06EA 55.29 USD InStock
AAB23836-06 AAB23836-14 CAAAB23836-22 CAAAB23836-14 CAAAB23836-06 AAB23836-22
3-Cyanobenzoic acid 98+%
3-Cyanobenzoic acid


Formula: C₈H₅NO₂
MW: 147.13 g/mol
Melting Pt: 219…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00002486
CAS Number: 1877-72-1
EINECS: 217-511-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty