Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ergosterol hydrate, (max. 6% H₂O) 96% (dry weight)

Supplier: Thermo Scientific Chemicals
Synonyms: 3β-Hydroxy-5,7,22-ergostatriene, 5,7,22-Ergostatrien-3β-ol hydrateProvitamin D2

B23840-14 B23840-14 B23840-06 B23840-06
AAB23840-06EA 72.39 USD InStock
AAB23840-06 CAAAB23840-06 AAB23840-14 CAAAB23840-14
Ergosterol hydrate, (max. 6% H₂O) 96% (dry weight)
Ergosterol hydrate
A membrane lipid found almost exclusively in fungi. It is used as an indicator of living fungal biomass


Formula: C₂₈H₄₄O·xH₂O
MW: 396.65 g/mol (anhy)
Melting Pt: 154…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00003623
CAS Number: 57-87-4
EINECS: 200-352-7
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty