Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-methylpropane 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Isobutyl bromide

B23854-0B B23854-0B B23854-30 B23854-30
AAB23854-30EA 57.59 USD InStock
AAB23854-30 CAAAB23854-0B CAAAB23854-30 AAB23854-0B
1-Bromo-2-methylpropane 98+%
1-Bromo-2-methylpropane


Formula: C₄H₉Br
MW: 137.02 g/mol
Boiling Pt: 90…92 °C
Melting Pt: –118 °C
Density: 1.255
Flash Pt: 18 °C(64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000217
CAS Number: 78-77-3
EINECS: 201-141-2
UN: 2342
ADR: "3",II
Merck Index: 13,05152
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty