Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Phenylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

B23930-06 B23930-14 B23930-14 B23930-06
AAB23930-14EA 186.99 USD InStock
AAB23930-14 CAAAB23930-06 CAAAB23930-14 AAB23930-06
3-Phenylpyridine 97%
3-Phenylpyridine


Formula: C₁₁H₉N
MW: 155.2 g/mol
Boiling Pt: 272 °C
Melting Pt: 76…77 °C
Density: 1.085
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006380
CAS Number: 1008-88-4
EINECS: 213-762-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty