Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triacetoxymethylsilane 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Triacetoxy(methyl)silaneMethylsilanetriyl triacetate

B23974-09 B23974-18 B23974-18 B23974-09 B23974-30 B23974-30
AAB23974-30EA 506.29 USD InStock
AAB23974-30 AAB23974-09 CAAAB23974-30 AAB23974-18 CAAAB23974-09 CAAAB23974-18
Triacetoxymethylsilane 96%
Triacetoxymethylsilane


Formula: C₇H₁₂O₆Si
MW: 220.25 g/mol
Boiling Pt: 87…88 °C/3 mm
Melting Pt: 40…45 °C
Density: 1.175
Flash Pt: 85 °C(185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008694
CAS Number: 4253-34-3
EINECS: 224-221-9
UN: 1759
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty