Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methoxypropene 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Methoxy-1-propene

Stabilization: Stabilised with potassium carbonate ∼0.5%

B24083-30 B24083-18 B24083-18 B24083-30
AAB24083-30EA 147.99 USD InStock
CAAAB24083-30 AAB24083-30 CAAAB24083-18 AAB24083-18
2-Methoxypropene 95% stabilized
2-Methoxypropene


Formula: C₄H₈O
Boiling Pt: 35…36 °C
Density: 0.758
Flash Pt: -29 °C(-20 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014929
CAS Number: 116-11-0
EINECS: 204-125-3
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty