Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Piperazinone 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Piperazin-2-one

B24131-06 B24131-06 B24131-03 B24131-03
AAB24131-06EA 258.99 USD InStock
CAAAB24131-03 AAB24131-06 AAB24131-03 CAAAB24131-06
2-Piperazinone 97%
2-Piperazinone


Formula: C₄H₈N₂O
MW: 100.12 g/mol
Melting Pt: 129…134 °C
Storage Temperature: Ambient
MDL Number: MFCD01318687
CAS Number: 5625-67-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty