Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Trifluoromethyl)benzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

B24143-06 B24143-14 B24143-14 B24143-06
AAB24143-14EA 305.99 USD InStock
CAAAB24143-14 AAB24143-14 AAB24143-06 CAAAB24143-06
3-(Trifluoromethyl)benzylamine 97%
3-(Trifluoromethyl)benzylamine


Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 93…97 °C/28 mm
Density: 1.222
Flash Pt: 71 °C(159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008117
CAS Number: 2740-83-2
EINECS: 220-367-2
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty