Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-5-chloro-2-methoxybenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Metoclopramide Impurity C4-Amino-5-chloro-o-anisic acid

B24281-06 B24281-06 B24281-14 B24281-14
AAB24281-14EA 96.52 USD InStock
CAAAB24281-06 AAB24281-14 AAB24281-06 CAAAB24281-14
4-Amino-5-chloro-2-methoxybenzoic acid 98+%
4-Amino-5-chloro-2-methoxybenzoic acid


Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
Melting Pt: 205…208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00038794
CAS Number: 7206-70-4
EINECS: 230-582-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty