Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ω-Bromo-4-phenylacetophenone 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Biphenyl-4-yl bromomethyl ketone4-Phenylphenacyl bromide2-Bromo-4'-phenylacetophenoneα-Bromo-4-phenylacetophenoneBromomethyl p-phenylphenyl ketone

B24298-22 B24298-22 B24298-14 B24298-14 B24298-06 B24298-06
AAB24298-14EA 109.99 USD InStock
CAAAB24298-06 AAB24298-14 AAB24298-22 CAAAB24298-14 AAB24298-06 CAAAB24298-22
ω-Bromo-4-phenylacetophenone 97%
ω-Bromo-4-phenylacetophenone


Formula: C₁₄H₁₁BrO
MW: 275.14 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00000202
CAS Number: 135-73-9
EINECS: 205-217-6
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty