Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diaminomaleonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

B24328-14 B24328-22 B24328-14 B24328-22
AAB24328-22EA 80.89 USD InStock
AAB24328-22 AAB24328-14 CAAAB24328-22 CAAAB24328-14
Diaminomaleonitrile 98%
Diaminomaleonitrile


Formula: C₄H₄N₄
MW: 108.1 g/mol
Melting Pt: 178…182 °C (dec.)
Density: 1.360
Storage Temperature: Ambient
MDL Number: MFCD00001870
CAS Number: 1187-42-4
EINECS: 214-697-6
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty