Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: (2S,3R,4R,5R)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate

B24426-14 B24426-14 B24426-06 B24426-22 B24426-22 B24426-06
AAB24426-22EA 437.99 USD InStock
CAAAB24426-06 CAAAB24426-14 AAB24426-22 AAB24426-14 CAAAB24426-22 AAB24426-06
β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98%
beta-D-Ribofuranose-1,2,3,5-tetraacetate


Formula: C₁₃H₁₈O₉
MW: 318.28 g/mol
Melting Pt: 80…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00005358
CAS Number: 13035-61-5
EINECS: 235-898-5
Order Now
Specification Test Results
Optical rotation -13+/-0.5° (c=2.4 in chloroform)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty