Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chlorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (3-Chlorophenyl)metaboric acid3-Chlorobenzeneboronic acidDihydroxy(3-chlorophenyl)borane

B24444-06 B24444-14 B24444-03 B24444-06 B24444-03 B24444-14
AAB24444-06EA 71.09 USD InStock
CAAAB24444-03 CAAAB24444-14 CAAAB24444-06 AAB24444-06 AAB24444-14 AAB24444-03
3-Chlorophenylboronic acid 97%
3-Chlorophenylboronic acid


Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 180…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00161354
CAS Number: 63503-60-6
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty