Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isobutyryl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

B24472-22 B24472-36 B24472-36 B24472-22
AAB24472-36EA 109.54 USD InStock
AAB24472-36 AAB24472-22 CAAAB24472-36 CAAAB24472-22
Isobutyryl chloride 98%
Isobutyryl chloride


Formula: C₄H₇ClO
MW: 106.55 g/mol
Boiling Pt: 91…93 °C
Melting Pt: ca …90 °C
Density: 1.017
Flash Pt: 8 °C(46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000717
CAS Number: 79-30-1
EINECS: 201-194-1
UN: 2395
ADR: "3",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty