Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-2,4-Diamino-4-oxobutanoic acid hydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-D-Asn-OH·H₂OH-D-Asn-OH(R)-2-Aminosuccinamic acid hydrateD-Asparagine monohydrate

B24556-14 B24556-14 B24556-22 B24556-22
AAB24556-14EA 41.27 USD InStock
AAB24556-14 AAB24556-22 CAAAB24556-14 CAAAB24556-22
(R)-2,4-Diamino-4-oxobutanoic acid hydrate 99%
(R)-2,4-Diamino-4-oxobutanoic acid hydrate


Formula: C₄H₁₀N₂O₄
MW: 150.14 g/mol (132.12 anhy)
Melting Pt: 274…276 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149558
CAS Number: 5794-24-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty