Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-methylbenzenesulfonic acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Aminotoluene-3-sulfonic acid4-Aminotoluene-3-sulphonic acid

B24560-30 B24560-30
AAB24560-30EA 56.69 USD InStock
CAAAB24560-30 AAB24560-30
2-Amino-5-methylbenzenesulfonic acid 99%
2-Amino-5-methylbenzenesulphonic acid


Formula: C₇H₉NO₃S
MW: 187.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007908
CAS Number: 88-44-8
EINECS: 201-831-3
UN: 2585
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty