Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminopyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Pyrimidin-2-aminePyrimidin-2-ylamine

B24594-36 B24594-36 B24594-22 B24594-22
AAB24594-36EA 140.99 USD InStock
AAB24594-36 AAB24594-22 CAAAB24594-36 CAAAB24594-22
2-Aminopyrimidine 98%
2-Aminopyrimidine


Formula: C₄H₅N₃
MW: 95.1 g/mol
Boiling Pt: 159 °C/186 mm
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00006089
CAS Number: 109-12-6
EINECS: 203-648-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty