Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methylbenzo[b]thiophene-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B24624-03 B24624-06 B24624-14 B24624-06 B24624-03 B24624-14
AAB24624-03EA 53.35 USD InStock
CAAAB24624-03 CAAAB24624-14 AAB24624-03 AAB24624-14 AAB24624-06 CAAAB24624-06
3-Methylbenzo[b]thiophene-2-carboxylic acid 97%
3-Methylbenzo[b]thiophene-2-carboxylic acid


Formula: C₁₀H₈O₂S
MW: 192.24 g/mol
Melting Pt: 241…244 °C
Storage Temperature: Ambient
MDL Number: MFCD01830305
CAS Number: 3133-78-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty