Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromobenzylamine 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (3-Bromophenyl)methanamine

B24963-14 B24963-22 B24963-06 B24963-22 B24963-14 B24963-06
AAB24963-14EA 172.99 USD InStock
AAB24963-14 AAB24963-22 CAAAB24963-14 CAAAB24963-06 CAAAB24963-22 AAB24963-06
3-Bromobenzylamine 95%
3-Bromobenzylamine


Formula: C₇H₈BrN
MW: 186.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01026119
CAS Number: 10269-01-9
EINECS: 679-024-4
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty