Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Boc-α-phosphonoglycine trimethyl ester 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate

B25421-14 B25421-03 B25421-06
AAB25421-06EA 203.99 USD InStock
AAB25421-06 AAB25421-14 AAB25421-03
(±)-Boc-α-phosphonoglycine trimethyl ester 95%
(±)-Boc-α-phosphonoglycine trimethyl ester


Formula: C₁₀H₂₀NO₇P
MW: 297.24 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD02178893
CAS Number: 89524-98-1
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty