Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(4-Methoxyphenyl)imidazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-(4-Methoxyphenyl)-1H-imidazole

B25621-06 B25621-03
AAB25621-06EA 122.99 USD InStock
AAB25621-06 AAB25621-03
1-(4-Methoxyphenyl)imidazole 98%
1-(4-Methoxyphenyl)imidazole


Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
Boiling Pt: 120 °C/0.1 mm
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00060492
CAS Number: 10040-95-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty