Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile

H25844-06 H25844-14
AAH25844-06EA 82 USD InStock
AAH25844-06 AAH25844-14
2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile 98%
2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile


Formula: C₁₀H₈F₃NO
MW: 215.18 g/mol
Boiling Pt: 76…82 °C/12 mm
Density: 1.152
Flash Pt: 89 °C(152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001844
CAS Number: 80866-87-1
EINECS: 279-591-4
UN: 3276
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty