Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorobis(4-methylphenyl)phosphine 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Chlorodi(p-tolyl)phosphine

H25889-03 H25889-06
AAH25889-03EA 129.84 USD InStock
AAH25889-03 AAH25889-06
Chlorobis(4-methylphenyl)phosphine 95%
Chlorobis(4-methylphenyl)phosphine


Formula: C₁₄H₁₄ClP
Boiling Pt: 180…184 °C/10 mm
Density: 1.159
Storage Temperature: Ambient
MDL Number: MFCD01630844
CAS Number: 1019-71-2
EINECS: 000-000-0
UN: 1760
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty