Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Phenylalanine methyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-Methyl 2-amino-3-phenylpropanoate hydrochlorideH-D-Phe-OMe.HCl

H25920-06 H25920-14
AAH25920-06EA 49.89 USD InStock
AAH25920-06 AAH25920-14
D-Phenylalanine methyl ester hydrochloride 99%
D-Phenylalanine methyl ester hydrochloride


Formula: C₁₀H₁₃NO₂
MW: 215.68 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00066112
CAS Number: 13033-84-6
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty