Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (1S,2R)-1,2,3,6-Tetrahydrophthalic acid 2-hydrogen 1-methyl ester2-Hydrogen 1-Methyl (1S,2R)-1,2,3,6-tetrahydrophthalate(1S,2R)-1-Methyl cis-4-cyclohexene-1,2-dicarboxylate(1S,2R)-1-Methyl cis-1,2,3,6-tetrahydrophthalate(1R,6S)-6-(Methoxycarbonyl)cyclohex-3-enecarboxylic acid

H25950-03 H25950-06
AAH25950-03EA 97.39 USD InStock
AAH25950-03 AAH25950-06
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester 98%
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester is used in organic synthesis as a starting material and intermediate.


Formula: C₉H₁₂O₄
MW: 184.19 g/mol
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00075490
CAS Number: 88335-93-7
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty