Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluoro-2-methoxybenzyl bromide 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(Bromomethyl)-4-fluoro-1-methoxybenzene

H25957-14 H25957-06 H25957-03
AAH25957-14EA 405.77 USD InStock
AAH25957-14 AAH25957-03 AAH25957-06
5-Fluoro-2-methoxybenzyl bromide 98+%
5-Fluoro-2-methoxybenzyl bromide


Formula: C₈H₈BrFO
MW: 219.05 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00671769
CAS Number: 700381-18-6
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty