Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Pyrrolidinyl)pyridine 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Pyrrolidin-2-ylpyridine

H26051-03 H26051-03 H26051-MD
AAH26051-MDEA 49.19 USD InStock
CAAAH26051-03 AAH26051-MD AAH26051-03
4-(2-Pyrrolidinyl)pyridine 96%
4-(2-Pyrrolidinyl)pyridine


Formula: C₉H₁₂N₂
MW: 148.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862536
CAS Number: 128562-25-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty