Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(Trifluoromethyl)cyclopentanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H26055-03 H26055-MD
AAH26055-03EA 463.99 USD InStock
AAH26055-03 AAH26055-MD
1-(Trifluoromethyl)cyclopentanecarboxylic acid 97%
1-(Trifluoromethyl)cyclopentanecarboxylic acid


Formula: C₇H₉F₃O₂
MW: 182.15
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD08445821
CAS Number: 277756-44-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty