Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-4-methoxybenzyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-(Bromomethyl)-2-fluoro-1-methoxybenzene

H26116-03 H26116-MD
AAH26116-MDEA 86.39 USD InStock
AAH26116-MD AAH26116-03
3-Fluoro-4-methoxybenzyl bromide 98%
3-Fluoro-4-methoxybenzyl bromide


Formula: C₈H₈BrFO
MW: 219.05 g/mol
Melting Pt: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00671768
CAS Number: 331-61-3
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty