Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2-fluorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H26187-06 H26187-03 H26187-14
AAH26187-03EA 68.09 USD InStock
AAH26187-03 AAH26187-14 AAH26187-06
5-Chloro-2-fluorobenzaldehyde 97%
5-Chloro-2-fluorobenzaldehyde


Formula: C₇H₄ClFO
Melting Pt: 29…32 °C
Storage Temperature: Ambient
MDL Number: MFCD03094518
CAS Number: 96515-79-6
EINECS: None
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty