Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-4-nitrobenzyl bromide 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-(Bromomethyl)-2-fluoro-1-nitrobenzene

H26193-06 H26193-03
AAH26193-03EA 183.99 USD InStock
AAH26193-03 AAH26193-06
3-Fluoro-4-nitrobenzyl bromide 97%
3-Fluoro-4-nitrobenzyl bromide


Formula: C₇H₅BrFNO₂
MW: 234.024 g/mol
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD03094237
CAS Number: 131858-37-2
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty