Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-4-hydroxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H26218-ME H26218-MC
AAH26218-MCEA 116.99 USD InStock
AAH26218-MC AAH26218-ME
2-Fluoro-4-hydroxybenzaldehyde 97%
2-Fluoro-4-hydroxybenzaldehyde


Formula: C₇H₅FO₂
MW: 140.11 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD06797918
CAS Number: 348-27-6
EINECS: None
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty