Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Fluoro-4-ethynylbenzene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (4-Fluorophenyl)ethyne1-Ethynyl-4-fluorobenzene(4-Fluorophenyl)acetylene

H26269-06 H26269-06 H26269-14 H26269-03 H26269-03
AAH26269-14EA 780.99 USD InStock
AAH26269-14 AAH26269-03 CAAAH26269-06 CAAAH26269-03 AAH26269-06
1-Fluoro-4-ethynylbenzene 99%
1-Fluoro-4-ethynylbenzene


Formula: C₈H₅F
MW: 120.13 g/mol
Boiling Pt: 55…56 °C/40 mm
Melting Pt: 27…28 °C
Density: 1.048
Flash Pt: 29 °C(85 °F)
Storage Temperature: Ambient
MDL Number: MFCD00168823
CAS Number: 766-98-3
UN: 1325
ADR: "4.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty