Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Trifluoromethyl)thiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H26311-03 H26311-06 H26311-MD
AAH26311-03EA 68.89 USD InStock
AAH26311-03 AAH26311-06 AAH26311-MD
3-(Trifluoromethyl)thiobenzamide 97%
3-(Trifluoromethyl)thiobenzamide


Formula: C₈H₆F₃NS
MW: 151.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01567258
CAS Number: 53515-17-6
EINECS: None
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty