Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-5-fluoroaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

H26338-03 H26338-06
AAH26338-03EA 165.99 USD InStock
AAH26338-03 AAH26338-06
3-Chloro-5-fluoroaniline 97%
3-Chloro-5-fluoroaniline


Formula: C₆H₅ClFN
MW: 145.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407962
CAS Number: 4863-91-6
EINECS: None
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty