Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(4-(Trifluoromethyl)phenyl)methanesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Trifluoromethyl-α-toluenesulphonyl chloride[4-(Trifluoromethyl)phenyl]methanesulphonyl chloride[4-(Trifluoromethyl)phenyl]methanesulfonyl chloride4-Trifluoromethyl-α-toluenesulfonyl chloride

H26395-MD H26395-03
AAH26395-MDEA 51.38 USD InStock
AAH26395-MD AAH26395-03
(4-(Trifluoromethyl)phenyl)methanesulfonyl chloride 97%
(4-(Trifluoromethyl)phenyl)methanesulphonyl chloride


Formula: C₈H₆ClF₃O₂S
MW: 258.65 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
EINECS: None
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty