Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-L-prolinal 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-1-Boc-2-Formylpyrrolidine

H26495-06 H26495-03
AAH26495-03EA 149.99 USD InStock
AAH26495-03 AAH26495-06
N-Boc-L-prolinal 96%
N-Boc-L-prolinal


Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Boiling Pt: 228 °C
Density: 1.06
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00274186
CAS Number: 69610-41-9
Order Now
Specification Test Results
Optical rotation -95° (c=1, chloroform)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty