Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Formyl-6-methoxypyridine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Methoxy-3-pyridinecarboxaldehyde2-Methoxypyridine-5-carboxaldehyde6-Methoxynicotinaldehyde6-Methoxypyridine-3-carboxaldehyde

H26515-06 H26515-03
AAH26515-06EA 254.99 USD InStock
AAH26515-06 AAH26515-03
3-Formyl-6-methoxypyridine 98%
3-Formyl-6-methoxypyridine


Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 49…52 °C
Flash Pt: 107 °C(225 °F)
Storage Temperature: Ambient
MDL Number: MFCD02683446
CAS Number: 65873-72-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty