Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-2,6-difluorobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H26556-06 H26556-03
AAH26556-03EA 170.99 USD InStock
AAH26556-03 AAH26556-06
4-Chloro-2,6-difluorobenzoic acid 97%
4-Chloro-2,6-difluorobenzoic acid


Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
Melting Pt: 183…186 °C
Storage Temperature: Ambient
MDL Number: MFCD06203570
CAS Number: 196194-58-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty