Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

H26569-14 H26569-06
AAH26569-14EA 160.99 USD InStock
AAH26569-14 AAH26569-06
4-Pentylaniline 98%
4-Pentylaniline


Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 130 °C
Density: 0.919
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007926
CAS Number: 33228-44-3
EINECS: 251-409-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty