Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hexylfuran 97%

Supplier: Thermo Scientific Chemicals
Warning

H26698-03 H26698-14 H26698-06
AAH26698-03EA 45.71 USD InStock
AAH26698-03 AAH26698-14 AAH26698-06
2-Hexylfuran 97%
2-Hexylfuran


Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053134
CAS Number: 3777-70-6
EINECS: 223-235-2
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty