Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propionylfuran 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-(Furan-2-yl)propan-1-one

H26754-14 H26754-06
AAH26754-06EA 79.69 USD InStock
AAH26754-06 AAH26754-14
2-Propionylfuran 97%
2-Propionylfuran


Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 53…55 °C/1 mm
Melting Pt: 26…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00020936
CAS Number: 3194-15-8
EINECS: 221-693-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty