Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-5-(trifluoromethoxy)benzyl bromide 97%

Supplier: Thermo Scientific Chemicals
Danger

H26795-03 H26795-06
AAH26795-03EA 131.99 USD InStock
AAH26795-03 AAH26795-06
2-Fluoro-5-(trifluoromethoxy)benzyl bromide 97%
2-Fluoro-5-(trifluoromethoxy)benzyl bromide


Formula: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD04115890
CAS Number: 86256-24-8
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty