Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Fluoroisatin 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 7-Fluoroindoline-2,3-dione7-Fluoro-1H-indole-2,3-dione

H26827-14 H26827-06 H26827-03
AAH26827-14EA 283.99 USD InStock
AAH26827-14 AAH26827-03 AAH26827-06
7-Fluoroisatin 97%
7-Fluoroisatin


Formula: C₈H₄FNO₂
MW: 165.12 g/mol
Melting Pt: 193…195 °C
Storage Temperature: Ambient
MDL Number: MFCD01569508
CAS Number: 317-20-4
EINECS: None
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty