Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate 94%

Supplier: Thermo Scientific Chemicals
Danger

H26857-06 H26857-03 H26857-14 H26857-03
AAH26857-06EA 76.39 USD InStock
AAH26857-06 AAH26857-03 AAH26857-14 CAAAH26857-03
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate 94%
Trimethylsilyl 2,2-difluoro-2-(fluorosulphonyl)acetate


Formula: C₅H₉F₃O₄SSi
MW: 250.27 g/mol
Boiling Pt: 156 °C
Density: 1.27
Flash Pt: 26 °C(79 °F)
Storage Temperature: Ambient
MDL Number: MFCD02093343
CAS Number: 120801-75-4
UN: 2920
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty